Neutralrot, zertifizierbare Qualität
Lieferant: VWR Chemicals
Synonyme:
Basisrot 5
, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid
, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
E895-100GEA
964
EUR
E895-100G
Neutralrot, zertifizierbare Qualität
Neutralrot
A cationic azine dye slightly soluble in water and ethanol. Used as a weak nuclear stain and as a supravital stain of fresh peripheal blood.
Formel:
C₁₅H₁₇ClN₄ Molecular Weight: 288,78 g/mol Schmelzpunkt: 290 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012651 CAS-Nummer: 553-24-2 EINECS: 209-035-8 Merck Index: 12,06571 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...