Human thrombin (from plasma)
Lieferant: Calbiochem
605206-100EA 127 EUR
605206-100
Human thrombin (from plasma)
Enzyme
Native thrombin from human plasma. Formulation useful for procedures in which the presence of citrate is undesirable.
  • Citrate free