4-Brom-3,5-dimethylisoxazol

Lieferant: Thermo Scientific

MAYBCC00610.10EA 0 EUR
MAYBCC00610.10 MAYBCC00610DA
4-Brom-3,5-dimethylisoxazol
4-Bromo-3,5-dimethylisoxazole
Formel: C₅H₆BrNO
Molecular Weight: 176,01 g/mol
MDL: MFCD00068187
CAS-Nummer: 10558-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR