1-(4-Bromphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM LTD (CHEMPUR)
|
Gefahr
|
Synonyme:
N-(4-Bromphenyl)thioharnstoff
, 4-Bromphenylthioharnstoff
FLUO010660-5GEA
41.3
EUR
FLUO010660-5G
1-(4-Bromphenyl)-2-thioharnstoff
1-(4-Bromphenyl)-2-thioharnstoff
Formel:
(C₆H₄Br)NHC(S)NH₂ Molecular Weight: 231,12 g/mol Siedepunkt: 319 °C (1013 hPa) Schmelzpunkt: 186 °C |
MDL:
MFCD00041176 CAS-Nummer: 2646-30-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...