meso-Tetraphenylporphin

Lieferant: FLUOROCHEM LTD (CHEMPUR)

Synonyme: 5,10,15,20-Tetraphenyl-21H,23H-porphin

FLUO011221-10GEA 0 EUR
FLUO011221-10G
meso-Tetraphenylporphin
meso-Tetraphenylporphin
Formel: C₄₄H₃₀N₄
Molecular Weight: 614,75 g/mol
Dichte: 1,276 g/cm³ (20 °C)
MDL: MFCD00011680
CAS-Nummer: 917-23-7
EINECS: 213-025-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR