4-Nitrophthalimid

Lieferant: Molekula

Synonyme: 4-Nitrophthalsäureimid4-Nitro-1,2-benzoldicarbonsäureimid

MOLE67659894-25GEA 0 EUR
MOLE67659894-25G
4-Nitrophthalimid
4-Nitrophthalimid
Formel: C₈H₄N₂O₄
Molecular Weight: 192,13 g/mol
Schmelzpunkt: 215 °C
MDL: MFCD00005884
CAS-Nummer: 89-40-7
EINECS: 201-905-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR