Wasser nukleasefrei

Synonyme: Wasserstoffoxid, Hydrogenoxid

129115 129117
QIAG129115EA 121 EUR
QIAG129115 QIAG129117
Wasser nukleasefrei
Wasser

Nuclease-free water for use in any molecular biology application.


  • Highly pure
  • Nuclease-free and deionized without the use of chemical additives


Ideal for use in any molecular biology application. Includes nuclease-free water prepared without the use of diethylpyrocarbonate (DEPC).

Formel: H₂O
Molecular Weight: 18,02 g/mol
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR