4-Chlorbenzhydrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL124982-1GEA 44.7 EUR
SIAL124982-1G
4-Chlorbenzhydrol, Sigma-Aldrich®
4-Chlorbenzhydrol
Formel: C₁₃H₁₁ClO
Molecular Weight: 218,68 g/mol
MDL: MFCD00004491
CAS-Nummer: 119-56-2
EINECS: 204-333-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR