1-Butyl-3-methylimidazoliumchlorid
Lieferant: Strem Chemicals, Inc.
STRE07-0100-5GEA
0
EUR
STRE07-0100-5G
1-Butyl-3-methylimidazoliumchlorid
1-Butyl-3-methylimidazolium chloride
Formel:
C₈H₁₅ClN₂ Molecular Weight: 174,67 g/mol Siedepunkt: ∼200 °C (1013 hPa) Schmelzpunkt: ~ 60 °C Dichte: ∼1 g/cm³ (20 °C) Flammpunkt: >110 °C |
MDL:
MFCD03095425 CAS-Nummer: 79917-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...