Natriumhydrogensulfat

Lieferant: Bernd Kraft
Gefahr

Synonyme: Natriumbisulfat

KRAF22822.0600EA 55.7 EUR
KRAF22822.0600
Natriumhydrogensulfat
Natriumhydrogensulfat
Formel: HNaO₄S
Molecular Weight: 120,06 g/mol
MDL: MFCD00003531
CAS-Nummer: 7681-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR