uvex 3, MACSOLE®, 68631 ESD Safety Boots, Lace-Up
Lieferant: Uvex
Uvex 3 MACSOLE®
UVEX6863.1/45EA 196 EUR
UVEX6863.1/45
uvex 3, MACSOLE®, 68631 ESD Safety Boots, Lace-Up
Schuhe und Stiefel
The MACSOLE ® rubber outsole makes the companion even stronger for heavy-duty use - this makes the outsole particularly resistant to heat, chemicals and cuts. In addition, the MACSOLE® sole technology made of rubber impresses with unsurpassed slip resistance, even in the cold.
  • Hydrophobic leather upper material
  • Waterproof Sympatex lining with dust tab
  • Replaceable, antistatic comfort footbed
  • Heat-, chemical- and cut-resistant uvex MACSOLE® rubber outsole
  • Foamed polyurethane overcap and heel cup
  • 100% metal-free toe cap
  • Penetration resistance made of steel
  • Reflective elements
  • Uvex anklepro ankle protection
Robust, modern and lightweight safety lace-up boots with an extra wide fit. Dirt-resistant and smooth-running lacing system with uvex lacelock. Cushioning and energy-returning uvex i-PUREnrj midsole. Protection against electrostatic charging (ESD) with a leakage resistance of less than 35 megaohms.
Standards: EN ISO 20345:2022 S7 FO CI BY HRO SC SR; EN 61340-4-3 + EN 61340-5-1