DL-α-Phenylalanin

Lieferant: Molekula

Synonyme: 3-Phenyl-DL-alanin(±)-2-Amino-3-phenylpropionsäure(±)-2-Amino-3-phenylpropionic acid(±)-2-Amino-3-phenylpropansäureDL-PhenylalaninH-DL-Phe-OH

MOLEM31697582EA 0 EUR
MOLEM31697582
DL-α-Phenylalanin
DL-α-Phenylalanin
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: 173 °C
MDL: MFCD00064225
CAS-Nummer: 150-30-1
EINECS: 205-756-7
Merck Index: 12,07425

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR