Ficoll®
Lieferant: MP Biomedicals
Ficoll®
ICNA0216000380EA 533 EUR
ICNA0216000380
Ficoll®
Zelltrennmedien
A copolymer of sucrose and epichlorohydrin. Component used to make density gradients for lymphocyte separation.