VHG™ Optilene-XF™ XRF Thin Film
Lieferant: LGC Standards
VHG™
PROOVHG-FOL06-R3EA 185 EUR
PROOVHG-FOL06-R3
VHG™ Optilene-XF™ XRF Thin Film
Versiegeln Verschlussfolien
Thin films for XRF sample cups.
Highest grade, proprietary film featuring optimal transmission, purity, strength, chemical resistance, and thermal stability. Good for full range of XRF analytes, including light elements.