5,8,11,14-Eicosatetrainsäure ≥98%

Lieferant: ENZO LIFE SCIENCES AG

Synonyme: ETYAOctadehydroarachidonic acid

ENZOBMLET0040100EA 385 EUR
ENZOBMLET0040100 ENZOBMLET0040020
5,8,11,14-Eicosatetrainsäure ≥98%
5,8,11,14-Eicosatetraynoic acid

Arachidonic acid uptake inhibitor


Inhibits arachidonic acid uptake. Inhibits cyclooxygenase (ID50=8 µM) and all lipoxygenases (ID50=10, 0.3, and 0.2 µM for 5-, 12- and 15-LO respectively) in whole cells. Inhibits PLA2 and cytochrome P-450. Modulates Ca2+ entry into cells. Stimulates luteinizing hormone release from cultured pituitary cells. PPAR agonist.

Formel: C₂₀H₂₄O₂
MDL: MFCD00036967
CAS-Nummer: 1191-85-1
https://de.vwr.com/stibo/web/std.lang.all/38/32/45403832.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR