Streptomycinsulfat (aus Streptomyces species)

Lieferant: ENZO LIFE SCIENCES AG

ENZOALX380277G100EA 99 EUR
ENZOALX380277G100
Streptomycinsulfat (aus Streptomyces species)
Streptomycin sulphate

Antibiotic. Inhibits prokaryote protein synthesis. Binds to the S12 protein of the 30S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria. Mutation in rpsL (the gene for S12 ribosomal protein) prevents binding of streptomycin to ribosome. Inactivated by aminoglycoside phosphotransferase.


White to off-white powder


Soluble in water (50 mg/mL)


Verpackung: Shipping temperature: Ambient

Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Molecular Weight: 1457,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037023
CAS-Nummer: 3810-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR