Bromthymolblau 0,1% in Ethanol

Lieferant: Bernd Kraft
Gefahr

Synonyme: BTB3′,3′′-Dibromthymolsulfonphthalein

KRAF17883.3700EA 0 EUR
KRAF17883.3700
Bromthymolblau 0,1% in Ethanol
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
Molecular Weight: 624,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
UN: 1170
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR