Rhodamin 6G, rein
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid
ACRO191410500EA
51.9
EUR
ACRO191410500
Rhodamin 6G, rein
Rhodamin 6G
Formel:
C₂₈H₃₁ClN₂O₃ Molecular Weight: 479,02 g/mol Siedepunkt: ∼200 °C (1013 hPa) Schmelzpunkt: 229,4 °C |
MDL:
MFCD00012665 CAS-Nummer: 989-38-8 EINECS: 213-584-9 UN: 2811 |
Spezifikation Testergebnisse
Appearance | Red-brown to brown Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...