Rhodamin 6G, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid

SIAL83697-1GEA 117 EUR
SIAL83697-1G SIAL83697-250MG
Rhodamin 6G, Sigma-Aldrich®
Rhodamin 6G
Formel: C₂₈H₃₁ClN₂O₃
Molecular Weight: 479,02 g/mol
Siedepunkt: ∼200 °C (1013 hPa)
MDL: MFCD00012665
CAS-Nummer: 989-38-8
EINECS: 213-584-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR