Benzethoniumchlorid 0.004 mol/l

Lieferant: Bernd Kraft

Synonyme: Hyamine® 1622

KRAF04015.3000EA 0 EUR
KRAF04015.3000
Benzethoniumchlorid 0.004 mol/l
Benzethoniumchlorid
Formel: C₂₇H₄₂ClNO₂
Molecular Weight: 448,09 g/mol
MDL: MFCD00011742
CAS-Nummer: 121-54-0
EINECS: 204-479-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR