Riboflavin (Vitamin B2), Sigma-Aldrich®

Lieferant: Merck

Synonyme: LactoflavinVitamin B26,7-Dimethyl-9-D-ribitylisoalloxazin7,8-Dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-benzo[g]pteridin-2,4(3H,10H)-dionVitamin G(-)-Riboflavin

M-Clarity™ Qualitätsstufe = MQ200

SIALR7649-500GEA 959 EUR
SIALR7649-500G
Riboflavin (Vitamin B2), Sigma-Aldrich®
Riboflavin (Vitamin B2)
Formel: C₁₇H₂₀N₄O₆
Molecular Weight: 376,37 g/mol
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005022
CAS-Nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR