Hämatoxylin modifizierte Lösung (Harris), Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
Synonyme: HydroxybrasilinNaturschwarz 1
M-Clarity™ Qualitätsstufe = MQ500
HHS128-4LEA 357 EUR
HHS128-4L
Hämatoxylin modifizierte Lösung (Harris), Sigma-Aldrich®
Hämatoxylin-Färbelösung
General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.
  • 7 g/L certified hematoxylin
Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00078111
CAS-Nummer: 517-28-2
https://de.vwr.com/stibo/web/std.lang.all/01/33/26660133.jpg
Bestellen
Spezifikation Testergebnisse
Appearance (Colour) Purple to Dark Purple
Appearance (Form) Solution
Appearance (Turbidity) Clear
Performance Conforms
Nuclear chromatin purplish to blue and well defined
Expiration Date Period 18 months