Kristallviolett-Lösung, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Methylviolett 10B
, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid
, Basic Violet 3
, Methylrosanilin
, 4,4',4''-Tri(dimethylamino)trityliumchlorid
, Methylrosanilium
, Hexamethylen-p-rosanilinchlorid
, Methylrosaniliumchlorid
, Hexamethyl-p-rosaniliniumchlorid
, Hexamethylviolett
, blaues Pyoktanin
, CI 42555
, Gentianaviolett
M-Clarity™ Qualitätsstufe = MQ500
Kit component of Gram Stain kits HT90A and HT90T (for staining of heat-fixed films and tissue, respectively).
HT901 is composed of 2.3% certified crystal violet, 0.1% ammonium oxalate monohydrate, and 20% ethyl alcohol, SD3A.
Formel:
C₂₅H₃₀ClN₃ Molecular Weight: 407,99 g/mol Flammpunkt: 37,8 °C (geschlossener Tiegel) Lagertemperatur: Room Temperature |
MDL:
MFCD00011750 CAS-Nummer: 548-62-9 |
Spezifikation Testergebnisse
Appearance (Colour) | Purple/Violet |
Appearance (Form) | Solution |
Appearance (Turbidity) Essentially free of particles/precipitate |
Clear |
Performance | Conforms |
Gram positive organisms are purple-black in both bacteriology smears and tissue sections; Gram negative organisms are red in both bacteriology smears and tissue sections. | |
Expiration Date Period | 24 months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...