3-Noradamantanecarboxylic acid 97%

Lieferant: SIGMA-ALDRICH MICROSCOPY

276235-1GEA 308 EUR
276235-1G
3-Noradamantanecarboxylic acid 97%
3-Noradamantanecarboxylic acid

Building block for 'Anti-Bredt' strained molecules.

Formel: C₁₀H₁₄O₂
Molecular Weight: 166,22 g/mol
Schmelzpunkt: 105,..107 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00213463
CAS-Nummer: 16200-53-6

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Purity (Titration by NAOH) 96.5 - 103.5%
GC (area %) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR