Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine 90% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

452718-1GEA 126 EUR
452718-1G
Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine 90% (Farbstoffgehalt), Sigma-Aldrich®
Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine

Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine (ZnTPyP) forms an organic thin-film, which can be used to fabricate hybrid heterojunction solar cells. It is also used in the production of organic nanostructures, which find applications in organic field-effect transistors (OFET), organic light-emitting displays (OLED), and nanosensors.

Formel: C₄₀H₂₄N₈Zn
Molecular Weight: 682,06 g/mol
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00274658
CAS-Nummer: 31183-11-6

Bestellen

Spezifikation Testergebnisse

Appearance (Colour), Violet or Purple Conforms to Requirements
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength, In Chloroform (Ethanol Stabilised) 420- 426 nm
Extinction Coefficient ≥200,000
Carbon 62.3 - 79.2%
Nitrogen 14.5 - 18.5%
Purity (HPLC) ≥88.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR