Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Lieferant: Thermo Fisher Scientific
Savant™ SpeedVac™
531-1494EA 16087.5 EUR
531-1494 531-1495
Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Konzentratoren
Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ systems and kits.
  • Plug and play installation: Integrated solution includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit; no tubing connections needed
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
  • Audible alarm to alert user when operation is finished
  • Oil-free vacuum pump is virtually maintenance free
  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced vapor path from concentrator to refrigerated vapor trap accelerates evaporation
Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e. 120 × 1,5 ml tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e. 200 × 1,5 ml tubes, other formats).
Solvent and Solvent Combinations:
- Acetonitrile
- Ethanol
- Methanol
- Methylene Chloride
- Water
- Other organic solvents
Lieferumfang: SPD1030P1 Kit includes: Fully Integrated SpeedVac system, 40 × 1,5 ml rotor (RH40-11), CryoCool™ 1 L heat transfer fluid, glass condensation flask (GCF400).
SPD2030P1 Kit includes: Integrated SpeedVac system, 200 × 1,5 ml rotor (RH200-12), CryoCool™ 1 L heat transfer fluid, glass condensation flask (GCF400).