Extraction Thimbles
Lieferant: Sartorius
FT-1201-022100 FT-1201-025060 FT-1201-025070 FT-1201-028060 FT-1201-033060 FT-1201-033130 FT-1201-035150 FT-1201-040100 FT-1201-040123
SARTFT-1201-022100EA 113 EUR
SARTFT-1201-022100 SARTFT-1201-025060 SARTFT-1201-025070 SARTFT-1201-028060 SARTFT-1201-033060 SARTFT-1201-033130 537- SARTFT-1201-040100 SARTFT-1201-040123
Extraction Thimbles
Extraktoren Extraktionshülsen
Extraction thimbles are efficient solvent-based separation from solids, liquids, and gases.
  • Composed of high-purity cellulose and glass fiber for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Glass fiber thimbles offer remarkable resistance to high temperatures
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Extraction thimbles have been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. These include environmental
monitoring tasks such as the separation of particulates like dust and aerosols, as well as the analysis of gas or air streams. They are also indispensable in food quality control processes, where they assist in the extraction of substances such as fats, emulsifiers, and additives. Designed for versatility, our thimbles can be utilised with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
Free of binders and offer high purity. This ensures consistent and high overall flow rates. Cellulose can be used for very sensitive applications. Guarantee accuracy for any extraction system with common dimensions. Typical wall thicknesses for our C300 grade are 1,3 mm for inner diameters under 35 mm and 1,7 mm for inner diameters over 35 mm.
Applications: Primarily utilised in applied industrial processes, our thimbles are versatile consumables for various analytical procedures. They are particularly effective for measuring free lipid content in food and beverages, identifying additives in both food products and elastomers, and monitoring soil samples as well as hot air streams. Simply place the thimbles into your preferred extractor and employ the continuous extraction method for reliable and accurate results.