Digital Pressure Probes with Tube Port
Lieferant: Senseca Germany
GRSG486674EA 219 EUR
GRSG486674 GRSG486675 GRSG486676 GRSG486677 GRSG486678 GRSG486679
Digital Pressure Probes with Tube Port
Temperaturfühler
DX 210 and DX 240 are series of miniature digital absolute and differential pressure probes for the PRO D01/PRO D05 handheld meters.
  • Interchangeable
  • Low power
  • Flexible hose connector
  • Compact and rigid stainless steel body
  • Fast and accurate
  • Temperature compensated
  • Optional DAkkS/ACCREDIA certification available
The measurementis temperature compensated for enhanced accuracy. The probes are supplied factory-calibrated, and they are ready to use. The calibration data are stored in the probe, allowing for interchangeability without the need for recalibration. Equipped with 4-pin M12 connector 'DX-Sensor standard', connectable to PRO-D0x multifunctional meters.
Bestellinformation: They can be optionally supplied with a DAkkS/ACCREDIA calibration certificate.