di-Natrium-α-glycerophosphat Hydrat Gemisch aus Isomeren
Lieferant: EHRENSTORFER GMBH
Synonyme:
ortho-Phosphorsäure(2,3-dihydroxypropyl)ester Dinatriumsalz Hydrat
EHERTRC-G601603-1GEA
0
EUR
EHERTRC-G601603-1G
di-Natrium-α-glycerophosphat Hydrat Gemisch aus Isomeren
di-Natrium-α-glycerophosphat Hydrat
Formel:
C₃H₇Na₂O₆P·H₂O Molecular Weight: 216,04 g/mol (anhy) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00149082 CAS-Nummer: 55073-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...