Alternariol monomethyl ether

Lieferant: LGC Standards PROMOCHEM
Gefahr

PROOTRCA575770-5MGEA 672 EUR
PROOTRCA575770-5MG
Alternariol monomethyl ether
Alternariol monomethyl ether
Formel: C₁₅H₁₂O₅
Molecular Weight: 272,253 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00058532
CAS-Nummer: 23452-05-3
EINECS: 636-657-0
UN: 3462
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR