Nonyl-β-D-glucopyranosid

Lieferant: Molekula

Synonyme: Nonyl-β-D-glucosidNG

MOLEM85890168EA 0 EUR
MOLEM85890168
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306,4 g/mol
MDL: MFCD00063300
CAS-Nummer: 69984-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR