Polyvinylacetat, M.W. 35,000-45,000

Lieferant: VWR Chemicals

87121.290EA 0 EUR
87121.290
Polyvinylacetat, M.W. 35,000-45,000
Polyvinylacetat
Formel: [CH₂CH(O₂CCH₃)]n
MDL: MFCD00084457
CAS-Nummer: 9003-20-7

Bestellen

Spezifikation Testergebnisse

Viscosity (10 %; ethyl acetate) 2.5 - 3.0 mPa.s
Acidity Max. 0.008 meq/g
Residual monomer Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR