IPTG (Isopropyl-β-D-thiogalactopyranosid)

Lieferant: STRATAGENE EUROPE

Synonyme: IPTG

STRA300127EA 165 EUR
STRA300127
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel: C₉H₁₈O₅S
Molecular Weight: 238,31 g/mol
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 109,5…110.5 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR