Sudan IV

Lieferant: EHRENSTORFER GMBH
Achtung

Synonyme: Scarlet Red ScharlachFat Red BOil Red IVScarlet RedScarlet RSudan® IV (Scharlachrot)1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphtholOil Red BB1-([2-Methyl-4-[(2-methylphenyl)diazenyl]phenyl]diazenyl)naphthalen-2-olSudan M Rot 380Oil RedBiebrich scarlet RLipid CrimsonSolvent Red 24

EHERC16986104EA 91 EUR
EHERC16986104
Sudan IV
Sudan IV
Formel: C₂₄H₂₀N₄O
Molecular Weight: 380,45 g/mol
Schmelzpunkt: 188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003893
CAS-Nummer: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR