Acrylnitril 99+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Cyanpropen

ACRO149631000EA 27.5 EUR
ACRO149631000 149630000. ACRO149630010 ACRO149630025 ACRO149630100
Acrylnitril 99+%
Acrylnitril
Formel: C₃H₃N
Molecular Weight: 53,06 g/mol
Siedepunkt: 77,3 °C (760 mmHg)
Schmelzpunkt: –83,5 °C
Dichte: 0,8 g/cm³
Flammpunkt: –0,2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001927
CAS-Nummer: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: 3,I
Merck Index: 13,00133

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3900 to 1.3920 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.0 %
Color scale ≤15 APHA
Stabilizer 35 to 45 ppm MEHQ
Non-volatile matter ≤100 ppm
pH 5.9 to 7.6 (5 % in water)
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Impurity ≤300 ppm Acetonitrile
Impurity ≤10 ppm Acrolein
Impurity ≤5 ppm Hydrocyanic acid
Impurity ≤10 ppm Methyl-vinyl-ketone
Impurity ≤100 ppm Oxazole
Acidity (CH3COOH) ≤20 ppm
Aldehyde (C2H4O) ≤50 ppm
Acetone ≤100 ppm
Peroxides ≤0.2 ppm (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR