Coomassie Brillantblau G 250

Lieferant: BIOMOL RESEARCH LABORATORIES

Synonyme: C.I. 42655Coomassie Blau G 250Brillantblau GHydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylen]-3-methylcyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium MononatriumsalzBrillantindocyanin GAcid Blue 90

BMOL03282-1EA 0 EUR
BMOL03282-1
Coomassie Brillantblau G 250
Coomassie Brillantblau G 250
Formel: C₄₇H₄₈N₃NaO₇S₂
Molecular Weight: 854,04 g/mol
Dichte: >1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00078482
CAS-Nummer: 6104-58-1
EINECS: 228-058-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR