Quarz, BCR® CRM 1400-5000 µm
Lieferant: Merck
Synonyme:
Silicium(IV)oxid
, Sand, weißer Quarz
, Siliciumdioxid
SIALBCR132-700GEA
250
EUR
SIALBCR132-700G
Quarz, BCR® CRM 1400-5000 µm
Quarz
Quartz (particle size) BCR® certified Reference Material, 1400-5000 particle size
Formel:
SiO₂ Molecular Weight: 60,08 g/mol Siedepunkt: 2230 °C (1013 hPa) Schmelzpunkt: 1427 °C Dichte: 2,622…2.645 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00217788 CAS-Nummer: 14808-60-7 EINECS: 238-878-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...