Quarz, BCR® CRM 1.20-20.00 μm

Lieferant: Merck

Synonyme: Silicium(IV)oxidSand, weißer QuarzSiliciumdioxid

SIALBCR070-10GEA 250 EUR
SIALBCR070-10G
Quarz, BCR® CRM 1.20-20.00 μm
Quarz

Quartz (particle size) BCR® certified Reference Material, 1.20-20.00 μm particle size

Formel: SiO₂
Molecular Weight: 60,08 g/mol
Siedepunkt: 2230 °C (1013 hPa)
Schmelzpunkt: 1427 °C
Dichte: 2,622…2.645 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00217788
CAS-Nummer: 14808-60-7
EINECS: 238-878-4
https://de.vwr.com/stibo/web/std.lang.all/05/83/16640583.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR