1,6-Diphenylhexan

Lieferant: Thermo Scientific

MAYBBTBG00196.250EA 0 EUR
MAYBBTBG00196.250 MAYBBTBG00196.1
1,6-Diphenylhexan
1,6-Diphenylhexan
Formel: (C₆H₅)CH₂CH₂CH₂CH₂CH₂CH₂(C₆H₅)
Molecular Weight: 238,37 g/mol
CAS-Nummer: 1087-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR