VWR® Kühlakkus
Lieferant: Avantor
VWR®
216-1168EA 214 EUR
216-1168 216-1169
VWR® Kühlakkus
Kühlakkus
Long lasting, high performance temperature protection.
  • Hard plastic bottle pack is filled with water
  • Can be refrigerated or frozen to keep products cold or frozen
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals, and biologicals
Diese robusten, mit Gel gefüllten Kühlakkus eignen sich zur Kühlung von Produkten in Kühlboxen, Kühltaschen oder Verpackungen. Das Gel sorgt für eine gleichbleibende und zuverlässige Kühlung.
  • Nicht toxisch und lebensmittelecht
  • Für starke Beanspruchung
  • Für rauen Transport geeignet
Tough, resuable bottle packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator, and are effective when used with insulated shipping containers. Phase changes at 0 °C and returns to liquid state after thawing.