The Bio-Link BLX crosslinker is an UV irradiation system, mainly dedicated to the linking of nucleic acid to membranes and elimination of PCR contamination.
Precise irradiation in either energy (Joules/cm²) or time (seconds)
Storage of last UV setting
Safety interlock door with UV blocking observation window