Crosslinkers, Bio-Link BLX
Lieferant: Vilber Lourmat
VILB611110811EA 0 EUR
VILB611110811
Crosslinkers, Bio-Link BLX
The Bio-Link BLX crosslinker is an UV irradiation system, mainly dedicated to the linking of nucleic acid to membranes and elimination of PCR contamination.
  • Precise irradiation in either energy (Joules/cm²) or time (seconds)
  • Storage of last UV setting
  • Safety interlock door with UV blocking observation window
  • Up to nine presets each for energy and time units