Lackmus

Lieferant: Molekula

MOLE37751194-25GEA 104 EUR
MOLE37751194-25G
Lackmus
Litmus
MDL: MFCD00131528
CAS-Nummer: 1393-92-6
EINECS: 215-739-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR