Xylencyanol FF
Lieferant: Molekula
Synonyme: Acid Blue 147Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium
MOLE27558266-25GEA 0 EUR
MOLE27558266-25G
Xylencyanol FF
Xylencyanol FF
Formel: C₂₅H₂₇N₂NaO₆S₂
Molecular Weight: 538,62 g/mol
MDL: MFCD00040657
CAS-Nummer: 2650-17-1
EINECS: 224-728-5