Xylencyanol FF

Lieferant: Serva
Achtung

Synonyme: Acid Blue 147Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium

SERA38505.01EA 0 EUR
SERA38505.01
Xylencyanol FF
Xylencyanol FF
Formel: C₂₅H₂₇N₂NaO₆S₂
Molecular Weight: 538,62 g/mol
Schmelzpunkt: 295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040657
CAS-Nummer: 2650-17-1
EINECS: 220-167-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR