Bor, Pulver, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
1.12070.0025
1.12070.0010
1.12070.0025EA
146
EUR
1.12070.0025
1.12070.0010
Bor, Pulver, Sigma-Aldrich®
Boron
Formel:
B Molecular Weight: 10,81 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134034 CAS-Nummer: 7440-42-8 EINECS: 231-151-2 UN: 3178 ADR: 4.1,III Merck Index: 13,01333 |
Spezifikation Testergebnisse
Solubility | (20 °C) practically insoluble |
Melting Point | 2075 °C |
Molar Mass | 10.81 g/mol |
Bulk Density | 200 - 300 kg/m3 |
Boiling Point | 4000 °C |
Density | 1.73 g/cm3 (15 °C) |
Ignition Point | 700 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...