n-Heptan-[D16] (98% D)
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
n-Heptane-[²H₁₆]
CIL-DLM-423-1
CIL-DLM-423-5
PROOCIL-DLM-423-1EA
289
EUR
PROOCIL-DLM-423-1
PROOCIL-DLM-423-5
n-Heptan-[D16] (98% D)
n-Heptan-[D16]
Formel:
D₃C(CD₂)₅CD₃ Molecular Weight: 116,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062744 CAS-Nummer: 33838-52-7 UN: 1206 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...