Anthracen-[D10]
Lieferant: EHRENSTORFER GMBH
Synonyme:
Paranaphthalin-D10
EHERC20520100EA
80
EUR
EHERC20520100
Anthracen-[D10]
Anthracen-[D10]
Formel:
C₁₄D₁₀ Molecular Weight: 188,15 g/mol Siedepunkt: 340 °C (1013 hPa) Lagertemperatur: Kühlschrank |
CAS-Nummer:
1719-06-8 EINECS: 217-004-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...