Aluminiumnitrat Nonahydrat
Lieferant: Honeywell Chemicals
|
Achtung
|
HONE11023.9025EA
0
EUR
HONE11023.9025
Aluminiumnitrat Nonahydrat
Aluminiumnitrat Nonahydrat
Formel:
Al(NO₃)₃·9H₂O Molecular Weight: 375,13 g/mol Schmelzpunkt: 73 °C Dichte: 1,72 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149132 CAS-Nummer: 7784-27-2 EINECS: 236-751-8 UN: 1438 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...