Bis(pentafluorphenyl)carbonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL539899-5GEA 111 EUR
SIAL539899-5G
Bis(pentafluorphenyl)carbonat, Sigma-Aldrich®
Bis(pentafluorphenyl)carbonat
Formel: C₁₃F₁₀O₃
Molecular Weight: 394,13 g/mol
MDL: MFCD00368353
CAS-Nummer: 59483-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR